Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-28A/IFN-lambda 2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1587
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1587 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1587 Supplier R&D Systems Supplier No. AF1587
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

IL-28A/IFN-lambda 2 Polyclonal specifically detects IL-28A/IFN-lambda 2 in Human samples. It is validated for Western Blot, ELISA, Neutralization, Immunocytochemistry.

Specifications

Antigen IL-28A/IFN-lambda 2
Applications Western Blot, ELISA, Neutralization, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, ELISA, Neutralization 0.3-1.5 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. Q8IZJ0
Gene Alias interleukin-28A, IFN-lambda 2, IL28A, IL-28A, interferon, lambda 2
Gene Symbols IFNL2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-28A/IFN-lambda 2 Val26-Val200 Accession # Q8IZJ0
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282616
Test Specificity Detects human IL-28A/IFN-lambda 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 60% cross-reactivity with human IL-28B/IFN-lambda 3 is observed. In direct ELISAs, approximately 30% cross-reactivity with recombinant human IL-29 and less than 5% cross-reactivity with recombinant mouse IL-28 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.