Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-28B/IFN-lambda 3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$214.00 - $538.00
Specifications
Antigen | IL-28B/IFN-lambda 3 |
---|---|
Clone | 567108 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB52593SP
![]() |
R&D Systems
MAB52593SP |
25 μg |
Each for $214.00
|
|
|||||
MB52593100
![]() |
R&D Systems
MAB52593100 |
100 μg |
Each for $538.00
|
|
|||||
Description
IL-28B/IFN-lambda 3 Monoclonal specifically detects IL-28B/IFN-lambda 3 in Human samples. It is validated for ELISA.Specifications
IL-28B/IFN-lambda 3 | |
ELISA | |
Monoclonal | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
interleukin-28B, IFN-lambda 3, IL28B, IL-28B, IL28C, interferon, lambda 3 | |
Chinese hamster ovary cell line CHO-derived recombinant human IL-28B/IFN-lambda 3, Arg30-Val200 (Lys74Arg), Accession No. Q81Z19 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IL-28B/IFN-lambda 3 in direct ELISAs. |
567108 | |
ELISA | |
Unconjugated | |
Human | |
Q81Z19 | |
282617.0 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title