Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human IL-28B/IFN-lambda 3 ELISA Kit (Colorimetric)
SDP

Catalog No. NB183412 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB183412 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB183412 Supplier Novus Biologicals™ Supplier No. NBP306788
This item is not returnable. View return policy

Colorimetric ELISA Kit

Specifications

Assay Range 15.63 to 1000 pg/mL
Assay Sensitivity 9.38 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Biological Fluids, Cell Culture Supernatant, Plasma, Serum
Specificity This kit recognizes Human IL-28B/IFN-lambda 3 in samples. No significant cross-reactivity or interference between Human IL-28B/IFN-lambda 3 and analogues was observed.
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 282617
Gene Symbol IFNL3
Quantity 1 Kit
Research Discipline Biologically Active Proteins
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym interleukin-28B, IL28B, IL-28B, IL28C, interferon, lambda 3
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.