Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-29/IFN-lambda 1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB15981SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB15981SP 25 μg
MAB15981100 100 μg
MAB15981500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB15981SP Supplier R&D Systems Supplier No. MAB15981SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 9 publications

IL-29/IFN-lambda 1 Monoclonal specifically detects IL-29/IFN-lambda 1 in Human samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen IL-29/IFN-lambda 1
Applications Neutralization, ELISA
Classification Monoclonal
Clone 247801
Concentration LYOPH
Conjugate Unconjugated
Dilution Neutralization 1-4 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8IU54
Gene Alias cytokine Zcyto21, IFNL1, IFNl3, IFNlambda 1, IFN-lambda 1, IFNlambda 3, IFN-lambda 3, IFN-lambda-1, IL29, IL-29, interferon lambda-1, interferon, lambda 1, interleukin 29 (interferon, lambda 1), interleukin-29, ZCYTO21
Gene Symbols IFNL1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-29/IFN-lambda 1 Gly20-Thr200 Accession # Q8IU54
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282618
Test Specificity Detects human IL-29/IFN-lambda 1 in ELISAs. In sandwich immunoassays, 100% cross-reactivity with recombinant human (rh) IL-28B/IFN-lambda 3 and 20% with rhIL-28A is observed. No cross-reactivity or interference was observed with rhIFN-alpha A, rhIFN-alpha B2, rhIFN-alpha G, rhIFN-alpha I, or rhIFN-beta 1A.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.