Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-29/IFN-lambda 1 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Supplier: R&D Systems BAF1598
This item is not returnable.
View return policy
Description
IL-29/IFN-lambda 1 Polyclonal specifically detects IL-29/IFN-lambda 1 in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
IL-29/IFN-lambda 1 | |
Polyclonal | |
Biotin | |
Q8IU54 | |
IFNL1 | |
Mouse myeloma cell line NS0-derived recombinant human IL-29/IFN-lambda 1 Gly20-Thr200 Accession # Q8IU54 | |
50 μg | |
Primary | |
Detects human IL-29/IFN-lambda 1 in ELISAs and Western blots. In sandwich immunoassays, approximately 100% cross-reactivity with recombinant human (rh) IL-28B is observed, 35% cross-reactivity with rhIL-28A is observed and less than 0.2% cross-reactivity with rhIFN-alpha A, rhIFN-alpha B2, rhIFN-alpha G, rhIFN alpha I, and rhIFN-beta 1A is observed. In Western blots, approximately 25% cross-reactivity with rhIL-28A and rhIL-28B is observed. | |
Human | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
cytokine Zcyto21, IFNL1, IFNl3, IFNlambda 1, IFN-lambda 1, IFNlambda 3, IFN-lambda 3, IFN-lambda-1, IL29, IL-29, interferon lambda-1, interferon, lambda 1, interleukin 29 (interferon, lambda 1), interleukin-29, ZCYTO21 | |
Goat | |
Affinity Purified | |
RUO | |
282618 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction