Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IL-29/IL-28B (IFN-lambda 1/3) DuoSet ELISA
SDP

Catalog No. DY1598B05 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY1598B05 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY1598B05 Supplier R&D Systems™ Supplier No. DY1598B05
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Human IL-29/IL-28B Capture Antibody; Human IL-29/IL-28B Detection Antibody; Human IL-29/IL-28B Standard; Streptavidin-HRP

Human IL-29/IL-28B (IFN-lambda 1/3) DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IL-29/IL-28B (IFN-lambda 1/3) DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IL-29/IL-28B IFN-lambda 1/3).

Specifications

Quantity 5 Plates
Species Human
Conjugate Biotin
For Use With (Application) ELISA
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.