Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-31 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$188.28
Specifications
| Antigen | IL-31 |
|---|---|
| Clone | 308202 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Applications | Western Blot, Immunocytochemistry |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB28241SP
![]() |
R&D Systems
MAB28241SP |
25 μg |
Each for $188.28
|
|
|||||
|
MAB28241
|
R&D Systems
MAB28241 |
100 μg | N/A | N/A | N/A | ||||
Description
IL-31 Monoclonal specifically detects IL-31 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
| IL-31 | |
| LYOPH | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q6EBC2 | |
| 386653 | |
| E. coli-derived recombinant human IL-31 Ser24-Thr164 Accession # Q6EBC2 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IL-31 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse IL-31 is observed. |
| 308202 | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL31, IL-31interleukin-31, interleukin 31 | |
| IL31 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title