Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IL-31, AOF Recombinant Protein

Catalog No. p-7203276
Encompass
Click to view available options
Quantity:
500 μg

E. coli Recombinant Protein

Interleukin-31 (IL-31) is a recently discovered T-cell cytokine closely related to IL-6 type cytokines and is preferentially produced by T helper type 2 cells. IL-31 activity is mediated through the ligand-induced oligomerization of a dimeric receptor complex containing IL-31 receptor A and oncostatin M receptor. In response to IL-31 binding, these proteins activate the JAK/STAT and the AKT signaling pathways. RNA levels of IL-31 receptor A and oncostatin M receptor are induced in activated monocytes but are expressed constitutively in epithelial cells. IL-31, when overexpressed in transgenic mice, results in the development of pruritis, alopecia, and skin lesions and in humans may result in atopic dermatitis, suggesting that IL-31 may represent a novel target for antipruritic drug development.
  • Purity: >95% by SDS-Page
  • Endotoxin Concentration: AX41 ≤EUs/μg
  • Protein Length: 141 aa.
  • Biological Activity: <25 ng/mL
  • Molecular Weight: 15.8 kDa
  • Animal Origin Free: Yes
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6EBC2
For Use With (Application) Control, ELISA, Functional Assay, Western Blot
Formulation 20 mM sodium phosphate with 50 mM NaCl and no preservative
Gene ID (Entrez) 386653
Molecular Weight (g/mol) 15.8 kDa
Name Human IL-31, AOF
Quantity 500 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Gene Alias 1700013B14Rik; Il31; IL-31; ILN; Interleukin; interleukin 31; Interleukin-31
Gene Symbol Il31
Specific Reactivity Human
Conjugate Unconjugated
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade >95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.