Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-31 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF2824
This item is not returnable.
View return policy
Description
IL-31 Polyclonal specifically detects IL-31 in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
IL-31 | |
Polyclonal | |
Biotin | |
NP_001014358 | |
IL31 | |
E. coli-derived recombinant human IL-31 (R&D Systems, Catalog # 2824-IL) Ser24-Thr164 Accession # NP_001014358 | |
50 μg | |
Primary | |
Detects human IL-31 in ELISAs and Western blots. In sandwich immunoassays, less than 1% cross-reactivity with recombinant mouse IL-31 is observed. | |
Human | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
IL31, IL-31interleukin-31, interleukin 31 | |
Goat | |
Affinity Purified | |
RUO | |
386653 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction