Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-31RA Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2769SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2769-SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB2769-SP Supplier R&D Systems Supplier No. MAB2769SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

IL-31RA Monoclonal specifically detects IL-31RA in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-31RA
Applications Flow Cytometry
Classification Monoclonal
Clone 313308
Conjugate Unconjugated
Dilution Flow Cytometry 0.25μg/106 cells
Formulation Lyophilized from a 0.2μm filtered solution in Tris, NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Gene Accession No. NP_620586
Gene Alias class I cytokine receptor, CRL3, CRL3Glmr, Cytokine receptor-like 3, GLMR, GLM-R, GLM-RMGC125346, GP130 like receptor, Gp130-like monocyte receptor, Gp130-like receptor, hGLM-R, IL-31 RA, IL-31 receptor subunit alpha, IL-31R subunit alpha, IL31RA, IL-31RAGPLCRL, IL-31R-alpha, interleukin 31 receptor A, interleukin-31 receptor subunit alpha, PRO21384, soluble type I cytokine receptor CRL3, ZcytoR17
Host Species Rat
Immunogen NS0-derived rhIL-31 RA extracellular domain aa 20-516 Accession # NP_620586
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 133396
Test Specificity Detects human IL-31 RA in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse IL-31 RA is observed.
Reconstitution Reconstitute at 0.5mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.