Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-31RA PE-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems FAB2769P
This item is not returnable.
View return policy
Description
IL-31RA Polyclonal specifically detects IL-31RA in Human samples. It is validated for Flow Cytometry.Specifications
IL-31RA | |
Polyclonal | |
Flow Cytometry 10 uL/10^6 cells | |
class I cytokine receptor, CRL3, CRL3Glmr, Cytokine receptor-like 3, GLMR, GLM-R, GLM-RMGC125346, GP130 like receptor, Gp130-like monocyte receptor, Gp130-like receptor, hGLM-R, IL-31 RA, IL-31 receptor subunit alpha, IL-31R subunit alpha, IL31RA, IL-31RAGPLCRL, IL-31R-alpha, interleukin 31 receptor A, interleukin-31 receptor subunit alpha, PRO21384, soluble type I cytokine receptor CRL3, ZcytoR17 | |
Goat | |
Affinity Purified | |
RUO | |
133396 | |
Human | |
IgG |
Flow Cytometry | |
R-PE | |
Q8NI17 | |
IL31RA | |
Mouse myeloma cell line NS0-derived recombinant human IL-31 RA Ala20-Ser516 Accession # Q8NI17 | |
100 Tests | |
Primary | |
Detects human IL-31 RA in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse IL-31 RA is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction