Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-32 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems IC30402V100UG
This item is not returnable.
View return policy
Description
IL-32 Monoclonal specifically detects IL-32 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
IL-32 | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
IL32, IL-32, IL-32alpha, IL-32beta, interleukin 32, interleukin-32, interleukin-32 theta, natural killer cell transcript 4, Natural killer cells protein 4, NK4, NK4IL-32delta, TAIF, TAIFa, TAIFb, TAIFc, TAIFd, TAIFIL-32gamma, Tumor necrosis factor alpha-inducing factor | |
E. coli-derived recombinant human IL-32 alpha Met1-Asn131 Accession # P24001 | |
100 μg | |
9235 | |
Human | |
Supernatant |
Flow Cytometry | |
373821 | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P24001 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human IL-32 alpha and other IL-32 isoforms in direct ELISAs. In direct ELISAs, 30% cross-reactivity with recombinant human (rh) IL-32 beta and 38% cross-reactivity with rhIL-32 gamma is detected. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction