Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-32 Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC30402R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC30402R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC30402R100 Supplier R&D Systems Supplier No. IC30402R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

IL-32 Monoclonal specifically detects IL-32 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen IL-32
Applications Flow Cytometry
Classification Monoclonal
Clone 373821
Conjugate Alexa Fluor 647
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P24001
Gene Alias IL32, IL-32, IL-32alpha, IL-32beta, interleukin 32, interleukin-32, interleukin-32 theta, natural killer cell transcript 4, Natural killer cells protein 4, NK4, NK4IL-32delta, TAIF, TAIFa, TAIFb, TAIFc, TAIFd, TAIFIL-32gamma, Tumor necrosis factor alpha-inducing factor
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-32 alpha Met1-Asn131 Accession # P24001
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 9235
Test Specificity Detects human IL-32 alpha and other IL-32 isoforms in direct ELISAs. In direct ELISAs, 30% cross-reactivity with recombinant human (rh) IL-32 beta and 38% cross-reactivity with rhIL-32 gamma is detected.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.