Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-32 alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB30401SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB30401SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB30401SP Supplier R&D Systems Supplier No. MAB30401SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 3 publications

IL-32 alpha Monoclonal specifically detects IL-32 alpha in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen IL-32 alpha
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 373802
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_001012651
Gene Alias IL32, IL32 alpha, IL-32alpha, IL-32beta, IL-32delta, IL-32gamma, interleukin 32, NK4, TAIF, TAIFa, TAIFb, TAIFc, TAIFd
Gene Symbols IL32
Host Species Rat
Immunogen E. coli-derived recombinant human IL-32 alpha Phe3-Lys131 Accession # NP_001012651
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9235
Test Specificity Detects the alpha isoform of IL-32 in direct ELISAs. In direct ELISAs, less than 15% cross-reactivity with recombinant human (rh) IL-32 beta and rhIL-32 gamma is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.