Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-33 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Goat Monoclonal Antibody
Supplier: R&D Systems IC36253S100UG
This item is not returnable.
View return policy
Description
IL-33 Monoclonal specifically detects IL-33 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
IL-33 | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
C9orf26, C9orf26chromosome 9 open reading frame 26 (NF-HEV), DKFZp586H0523, DVS27, DVS27-related protein, IL1F11, IL-1F11, IL33, IL-33, interleukin 33, Interleukin-1 family member 11, interleukin-33, NFHEV, NF-HEV, NF-HEVNFEHEV, Nuclear factor from high endothelial venules, RP11-575C20.2 | |
E.coli-derived recombinant human IL-33, Ser112-Thr270, Accession # O95760 | |
100 μg | |
90865 | |
Human | |
Supernatant |
Flow Cytometry | |
40015C | |
Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells | |
O95760 | |
Goat | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human IL-33 in direct ELISAs and Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction