Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-33 Antibody, R&D Systems™

Goat Monoclonal Antibody
Supplier: R&D Systems MAB36253100
This item is not returnable.
View return policy
Description
IL-33 Monoclonal specifically detects IL-33 in Human samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, ELISA Capture (Matched Antibody Pair).Specifications
IL-33 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C9orf26, C9orf26chromosome 9 open reading frame 26 (NF-HEV), DKFZp586H0523, DVS27, DVS27-related protein, IL1F11, IL-1F11, IL33, IL-33, interleukin 33, Interleukin-1 family member 11, interleukin-33, NFHEV, NF-HEV, NF-HEVNFEHEV, Nuclear factor from high endothelial venules, RP11-575C20.2 | |
Goat | |
Protein A or G purified from cell culture supernatant | |
RUO | |
90865 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
40015C | |
Western Blot 1 ug/mL, Immunohistochemistry 0.1-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
O95760 | |
IL33 | |
E.coli-derived recombinant human IL-33 Ser112-Thr270 Accession # O95760 | |
100 μg | |
Primary | |
Detects human IL-33 in direct ELISAs and Western blots. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction