Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IL-33 DuoSet ELISA
SDP

Catalog No. DY3625B05 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit (for 15 Plates)
1 Kit (for 5 Plates)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY3625B05 1 Kit (for 5 Plates)
DY3625B 1 Kit (for 15 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DY3625B05 Supplier R&D Systems™ Supplier No. DY3625B05

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols

Human IL-33 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IL-33 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IL-33.

Specifications

Description Fucose Detection
Product Type ELISA Kit
Quantity 1 Kit (for 5 Plates)
Species Human
Assay Range 23.40 to 1500 pg/mL
Cross Reactivity Not observed
Format 96-well Strip Plate
For Use With (Application) ELISA
Product Line R&D Systems
Sample Type Cell Culture Supernatant, Serum, Plasma
Kit Contents Capture antibody, Detection antibody, Recombinant standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Research Area Cytokines
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.