Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IL-33 High Sensitivity Magnetic Luminex™ Performance Assay

Catalog No. lbhs3625
Click to view available options
Quantity:
1 Kit
This item is not returnable. View return policy
This item is not returnable. View return policy

Magnetic bead-based multiplex assay for the Luminex™ MAGPIX™ platform

  • Magnetic format allows for easier wash steps
  • Undergoes similar validation testing as Quantikine™ ELISA Kits
  • Precise and Specific
  • Requires a small sample volume (<50 μL)
  • Run your assay in just 3-5 hours
  • Mass-calibrated standards for consistent results with every new lot of material
  • Enough reagents for one 96-well plate

Specifications

Product Type Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform
Quantity 1 Kit
Species Human
Binding Target IL-33
Bead Type Magnetic
Assay Sensitivity 1.06pg/mL
Assay Range 2.44 - 10000pg/mL (Serum, Heparin Plasma, EDTA Plasma), 2.44 - 2500pg/mL (Cell Culture Supernates)
Gene ID (Entrez) 90865
Gene Symbol IL33
Sample Type Serum, Plasma (EDTA, Heparin), Cell Culture Supernatant
Sample Volume Cell Culture Supernates (100μL),Serum (50μL),EDTA Plasma (50μL),Heparin Plasma (50μL)
Kit Contents
  • Microparticle Concentrate
  • Biotin-Antibody Concentrate
Content And Storage Store the unopened product at 2° to 8°C. Do not use past expiration date.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.