Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-34 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC5265T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC5265T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC5265T100 Supplier R&D Systems Supplier No. IC5265T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-34 Monoclonal specifically detects IL-34 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen IL-34
Applications Flow Cytometry
Classification Monoclonal
Clone 578416
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q6ZMJ4
Gene Alias C16orf77, IL34, IL-34, interleukin 34
Gene Symbols IL34
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-34 Asn21-Pro242 Accession # Q6ZMJ4
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146433
Test Specificity Detects recombinant human IL-34 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse IL-34 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.