Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-34 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC5265S100UG
This item is not returnable.
View return policy
Description
IL-34 Monoclonal specifically detects IL-34 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
IL-34 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q6ZMJ4 | |
IL34 | |
Mouse myeloma cell line NS0-derived recombinant human IL-34 Asn21-Pro242 Accession # Q6ZMJ4 | |
RUO | |
146433 | |
Human | |
IgG1 |
Flow Cytometry | |
578416 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
C16orf77, IL34, IL-34, interleukin 34 | |
Mouse | |
100 μg | |
Primary | |
Detects recombinant human IL-34 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse IL-34 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction