Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IL-34 Quantikine ELISA Kit

Catalog No. D3400
Click to view available options
Sufficient For:
1 x 96-well Plate
This item is not returnable. View return policy
This item is not returnable. View return policy

Human IL-34 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IL-34 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-34 in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul), saliva (50 ul), human milk (13 ul).

Specifications

Assay Range 31.30 - 2000pg/mL
Assay Sensitivity 3.06pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Human Milk, Plasma (EDTA, Heparin), Saliva, Serum
Specificity Natural and Recombinant Human IL-34
Sufficient For 1 x 96-well Plate
For Use With (Application) ELISA
Gene ID (Entrez) 146433
Sample Volume Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL),Saliva (50uL),Human Milk (13uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym C16orf77, IL34, IL-34, interleukin 34
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.