Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-36 alpha/IL-1F6 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
$214.00 - $585.00
Specifications
Antigen | IL-36 alpha/IL-1F6 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Description
IL-36 alpha/IL-1F6 Polyclonal specifically detects IL-36 alpha/IL-1F6 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
IL-36 alpha/IL-1F6 | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Human | |
FIL1, FIL1 epsilon, FIL1(EPSILON), FIL1E, IL-1 epsilon, IL1(EPSILON), IL1E, IL-1E, IL1F6, IL-1F6, IL-1F6 (FIL-1-epsilon), IL36 alpha, IL36A, interleukin 1 family, member 6 (epsilon), interleukin 1, epsilon, Interleukin 36, Alpha, Interleukin-1 epsilon, interleukin-1 family member 6, Interleukin-36 Alpha | |
IL36A | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
Q9UHA7 | |
27179 | |
E. coli-derived recombinant human IL-36 alpha/IL-1F6 Met1-Phe158 Accession # Q9UHA7 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human IL-36 alpha/IL-1F6 in direct ELISAs and Western blots. In Western blots, less than 2% cross-reactivity with recombinant human (rh) rhIL-36 beta , rhIL-36 gamma , rhIL-1 alpha , rhIL-1 beta and rhIL-18 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title