Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-36 beta/IL-1F8 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1099SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1099SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1099SP Supplier R&D Systems Supplier No. AF1099SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

IL-36 beta/IL-1F8 Polyclonal specifically detects IL-36 beta/IL-1F8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction, Neutralization.

Specifications

Antigen IL-36 beta/IL-1F8
Applications Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction 100 ug/mL, Neutralization 50-300 ng/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q3MIH0
Gene Alias family of interleukin 1-eta, FIL1, FIL1 eta, FIL1-(ETA), FIL1H, FILI-(ETA), IL-1 eta, IL1-ETA, IL1F8, IL-1F8, IL1F8 (Canonical product IL-1F8a), IL-1F8 (FIL1-eta), IL1H2, IL-1H2, IL36 beta, IL36B, interleukin 1 family, member 8 (eta), interleukin 1, eta, Interleukin 36, Beta, Interleukin-1 eta, interleukin-1 family member 8, Interleukin-1 homolog 2, Interleukin-1 Superfamily e, Interleukin-36 Beta
Gene Symbols IL36B
Host Species Goat
Immunogen E. coli-derived recombinant human IL-36 beta/IL-1F8 Met1-Glu157 Accession # Q3MIH0
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27177
Test Specificity Detects human IL-36 beta/IL-1F8 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) IL-36 gamma and rhIL-36 alpha is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.