Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IL-36 beta/IL-1F8 DuoSet ELISA
SDP

Catalog No. DY109905 Shop All Novus Biologicals Products
Change view
Click to view available options
Format:
96-well Strip Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
DY109905 96-well Strip Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY109905 Supplier R&D Systems™ Supplier No. DY109905

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols

For the determination of natural and recombinant Interleukin 36 beta in humans. R&D Systems Human IL-36 beta/IL-1F8 DuoSet ELISA Kit is a solid phase sandwich ELISA suitable for the analysis of cell culture supernate, serum and plasma samples.

The Human IL-36 beta/IL-1F8 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IL-36 beta/IL-1F8.

Specifications

Product Type ELISA Kit
Quantity 1 Kit (for 5 Plates)
Species Human
Assay Sensitivity 50 ng/mL
Cross Reactivity Not observed
Format 96-well Strip Plate
For Use With (Application) ELISA
Product Line R&D Systems
Sample Type Cell Culture Supernatant, Serum, Plasma
Kit Contents Capture antibody, Detection antibody, Recombinant standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Research Area Immunology
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.