Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-36 gamma/IL-1F9 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2320100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2320100 100 μg
MAB2320SP 25 μg
MAB2320500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB2320100 Supplier R&D Systems Supplier No. MAB2320100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 2 publications

IL-36 gamma/IL-1F9 Monoclonal specifically detects IL-36 gamma/IL-1F9 in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.

Specifications

Antigen IL-36 gamma/IL-1F9
Applications Western Blot, Blockade of Receptor-ligand Interaction
Classification Monoclonal
Clone 278706
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction 0.15-0.6 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9NZH8
Gene Alias IL-1 epsilon, IL-1 H1, IL-1 Related Protein 2, IL-1(EPSILON), IL1E, IL-1-epsilon, IL1F9, IL-1F9, IL1H1, IL-1H1, IL-1-Related Protein 2, IL1RP2, IL-1RP2, IL36 gamma, IL36G, interleukin 1 family, member 9, interleukin 1-related protein 2, Interleukin 36, Gamma, Interleukin-1 epsilon, interleukin-1 family member 9, Interleukin-1 homolog 1, Interleukin-36 Gamma
Gene Symbols IL36G
Host Species Rat
Immunogen E. coli-derived recombinant human IL-36 gamma/IL-1F9 Met1-Asp169 Accession # Q9NZH8
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56300
Test Specificity Detects human IL-36 gamma/IL-1F9 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) IL-1 alpha , rhIL-1 beta , rhIL-36Ra/IL-1F5, rhIL-36 alpha/IL-1F6, rhIL-1F7, rhIL-36 beta/IL-1F8, rhIL-1ra/IL-1F3, or rhIL-18/IL-1F4 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.