Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-37/IL-1F7 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
$200.50 - $2020.00
Specifications
Antigen | IL-37/IL-1F7 |
---|---|
Clone | 261506 |
Concentration | LYOPH |
Dilution | Blockade of Receptor-ligand Interaction 2-6 ug/mL |
Applications | Blockade of Receptor-ligand Interaction |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1975SP
![]() |
R&D Systems
MAB1975SP |
25 μg |
Each for $200.50
|
|
|||||
MAB1975100
![]() |
R&D Systems
MAB1975100 |
100 μg |
Each for $538.00
|
|
|||||
MAB1975500
![]() |
R&D Systems
MAB1975500 |
500 μg |
Each for $2,020.00
|
|
|||||
Description
IL-37/IL-1F7 Monoclonal specifically detects IL-37/IL-1F7 in Human samples. It is validated for Blockade of Receptor-ligand Interaction.Specifications
IL-37/IL-1F7 | |
LYOPH | |
Blockade of Receptor-ligand Interaction | |
Unconjugated | |
Mouse | |
Human | |
NP_775297 | |
27178 | |
E. coli-derived recombinant human IL-37/IL-1F7 Lys27-Asp192 Accession # NP_775297 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IL-37/IL-1F7 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-1 alpha , rhIL-1 beta , rhIL-1ra, rhIL-36Ra/IL-1F5, rhIL-36 alpha/IL-1F6, rhIL-36 beta/IL-1F8, or rhIL-18/IL-1F4 is observed. |
261506 | |
Blockade of Receptor-ligand Interaction 2-6 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FIL1, FIL1 zeta, FIL1(ZETA), FIL1ZFIL1 zeta, IL-1F7, IL-1F7IL-1 zeta, IL1H4IL1F7 (canonical product IL-1F7b), IL-1H4IL-1F7b (IL-1H4, IL-1H, IL-1RP1), IL-1RP1IL-1X protein, IL1RP1interleukin 1 family member 7, IL-1X, IL37, interleukin 1 family, member 7 (zeta), interleukin 1, zeta, interleukin-1 family member 7, Interleukin-1 homolog 4, interleukin-1 superfamily z, Interleukin-1 zeta, Interleukin-1-related protein | |
IL37 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title