Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-38/IL-1F10 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2427
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2427 100 μg
MAB2427SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2427 Supplier R&D Systems Supplier No. MAB2427
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

IL-38/IL-1F10 Monoclonal specifically detects IL-38/IL-1F10 in Human samples. It is validated for Western Blot.

Specifications

Antigen IL-38/IL-1F10
Applications Western Blot
Classification Monoclonal
Clone 316709
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8WWZ1
Gene Alias FIL1- theta, FKSG75, IL1F10, IL-1HY2, IL38, interleukin 1 family, member 10 (theta), UNQ6119/PRO20041
Gene Symbols IL1F10
Host Species Rat
Immunogen E. coli-derived recombinant human IL-38/IL-1F10 Met1-Trp152 Accession # Q8WWZ1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84639
Test Specificity Detects human IL-38/IL-1F10 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant mouse IL-1F10 is observed and no cross-reactivity with recombinant human (rh) IL-1 alpha/IL-1F1 rhIL-1 beta/IL-1F2, rhIL-1ra/IL-1F3, rhIL-18/IL-1F4, rhIL-36Ra/IL-1F5, rhIL-36 alpha/IL-1F6, rhIL-1F7/FIL1 zeta , rhIL-36 beta/IL-1F8, or rhIL-36 gamma/IL-1F9 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.