Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-38/IL-1F10 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB24271100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB24271100 100 μg
MAB24271SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB24271100 Supplier R&D Systems Supplier No. MAB24271100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-38/IL-1F10 Monoclonal specifically detects IL-38/IL-1F10 in Human samples. It is validated for ELISA.

Specifications

Antigen IL-38/IL-1F10
Applications ELISA
Classification Monoclonal
Clone 892257
Conjugate Unconjugated
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q8WWZ1
Gene Alias FIL1- theta, FKSG75, IL1F10, IL-1HY2, IL38, interleukin 1 family, member 10 (theta), UNQ6119/PRO20041
Host Species Mouse
Immunogen E. coli-derived human IL-38/IL-1F10, Ser3-Trp152, Accession # Q8WWZ1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84639
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.