Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-38/IL-1F10 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF2427
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF2427 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF2427 Supplier R&D Systems Supplier No. BAF2427
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

IL-38/IL-1F10 Polyclonal specifically detects IL-38/IL-1F10 in Human samples. It is validated for Western Blot.

Specifications

Antigen IL-38/IL-1F10
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q8WWZ1
Gene Alias FIL1- theta, FKSG75, IL1F10, IL-1HY2, IL38, interleukin 1 family, member 10 (theta), UNQ6119/PRO20041
Gene Symbols IL1F10
Host Species Goat
Immunogen E. coli-derived recombinant human IL-38/IL-1F10 Cys2-Trp152 Accession # Q8WWZ1
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84639
Test Specificity Detects human IL-38/IL-1F10 in Western blots. In Western blots, approximately 5% cross-reactivity with recombinant human (rh) IL-1ra and rhIL-1F5 is observed and less than 1% cross-reactivity with rhIL-1 alpha , rhIL-18, rhIL-1F6, rhIL-1F7, rhIL-1F8, and rhIL-1F9 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.