Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-3R alpha/CD123 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 7 publications
$214.00 - $2070.00
Specifications
Antigen | IL-3R alpha/CD123 |
---|---|
Clone | 32703 |
Dilution | Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, Neutralization 0.6-1.2 ug/mL, Immunocytochemistry 8-25 ug/mL, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOF |
Classification | Monoclonal |
Description
IL-3R alpha/CD123 Monoclonal specifically detects IL-3R alpha/CD123 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOF-ready.Specifications
IL-3R alpha/CD123 | |
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, Neutralization 0.6-1.2 ug/mL, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
Monoclonal | |
Mouse | |
Human | |
P26951 | |
3563 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-3 R alpha/CD123 Lys20-Arg305, predicted Accession # P26951 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IL-3 R alpha/CD123 in direct ELISAs and Western blots. |
32703 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOF | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 R alpha, IL-3 receptor subunit alpha, IL3R, IL3R alpha, IL-3R subunit alpha, IL3RA, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174 | |
IL3RA | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title