Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-4I1 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $467.50
Specifications
Antigen | IL-4I1 |
---|---|
Clone | 1006209 |
Applications | ELISA |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB56841SP
![]() |
R&D Systems
MAB56841SP |
25 μg |
Each for $200.50
|
|
|||||
MAB56841100
![]() |
R&D Systems
MAB56841100 |
100 μg |
Each for $467.50
|
|
|||||
Description
IL-4I1 Monoclonal specifically detects IL-4I1 in Human samples. It is validated for ELISA.Specifications
IL-4I1 | |
ELISA | |
Unconjugated | |
Rat | |
Human | |
Q96RQ9 | |
259307 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
1006209 | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
FIG1, Fig-1 protein, hFIG1, IL4I1, IL4-induced protein 1, interleukin 4 induced 1, interleukin four induced 1, LAAO, L-amino-acid oxidase, LAO, UNQ636/PRO1265 | |
Chinese hamster ovary cell line CHO-derived recombinant human IL-4I1, Met1-His567, Accession # Q96RQ9 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title