Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-4I1 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $503.00
Specifications
| Antigen | IL-4I1 |
|---|---|
| Clone | 1006202 |
| Dilution | Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 5-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB5684SP
![]() |
R&D Systems
MAB5684SP |
25 μg |
Each for $200.50
|
|
|||||
MAB5684100
![]() |
R&D Systems
MAB5684100 |
100 μg |
Each for $503.00
|
|
|||||
Description
IL-4I1 Monoclonal specifically detects IL-4I1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
| IL-4I1 | |
| Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 5-25 ug/mL | |
| Monoclonal | |
| Rat | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| FIG1, Fig-1 protein, hFIG1, IL4I1, IL4-induced protein 1, interleukin 4 induced 1, interleukin four induced 1, LAAO, L-amino-acid oxidase, LAO, UNQ636/PRO1265 | |
| Chinese Hamster Ovary cell line, CHO-derived human IL-4I1, Met1-His567, Accession No. Q96RQ9 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IL-4I1 in direct ELISAs. |
| 1006202 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Human | |
| Q96RQ9 | |
| 259307.0 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title