Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-5 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAM6051
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAM6051 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAM6051 Supplier R&D Systems Supplier No. BAM6051
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 3 publications

IL-5 Monoclonal specifically detects IL-5 in Human samples. It is validated for ELISA Detection (Matched Antibody Pair).

Specifications

Antigen IL-5
Applications ELISA
Classification Monoclonal
Clone JES1-5A10
Concentration LYOPH
Conjugate Biotin
Dilution ELISA Detection (Matched Antibody Pair) 0.5-2.0 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative
Gene Alias BCDF mu, B-cell differentiation factor I, BCGFII, EDF, Eo-CSF, Eosinophil differentiation factor, IL5, IL-5T-cell replacing factor, interleukin 5 (colony-stimulating factor, eosinophil), interleukin-5, TRF, TRFB cell differentiation factor I
Gene Symbols IL5
Host Species Rat
Immunogen COS-7 African green monkey SV40 transformed kidney fibroblast-like cell line-derived recombinant human IL-5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3567
Test Specificity Detects recombinant human IL-5 in ELISAs and Western blots. In sandwich immunoassays, no cross-reactivity with recombinant human (rh) IL-3, rhGM-CSF, or recombinant mouse IL-5 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.