Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-5 R alpha/CD125 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF253
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF253 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF253 Supplier R&D Systems Supplier No. BAF253
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

IL-5R alpha/CD125 Polyclonal specifically detects IL-5R alpha/CD125 in Human samples. It is validated for Western Blot.

Specifications

Antigen IL-5R alpha/CD125
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Alias CD125, CD125 antigen, CDw125CD125, HSIL5R3, IL-5 receptor subunit alpha, IL5R, IL5R alpha, IL-5R subunit alpha, IL5RA, IL-5RA, IL-5R-alpha, interleukin 5 receptor type 3, interleukin 5 receptor, alpha, interleukin-5 receptor alpha chain, interleukin-5 receptor alpha subunit, interleukin-5 receptor subunit alpha, MGC26560
Gene Symbols IL5RA
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-5 R alpha/CD125
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3568
Test Specificity Detects human IL-5 R alpha/CD125 in Western blots. In this format, less than 5% cross-reactivity with recombinant human (rh) IL-2 sR beta , rhIL-2 sR gamma , rhIL-3 sR, rhIL-4 sR, rhIL-1 sRII, rhIL-6 sR, rhIL-9 sR, and rhIL-10 sR is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.