Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-6 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC2061N100UG
This item is not returnable.
View return policy
Description
IL-6 Monoclonal specifically detects IL-6 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
IL-6 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A | |
E. coli-derived recombinant human IL-6 Val30-Met212 Accession # P05231 | |
100 μg | |
Primary | |
Detects human IL-6 in direct ELISAs. Does not cross-react with recombinant IL-6 from mouse, rat, or pig. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
Flow Cytometry | |
1936 | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P05231 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3569 | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction