Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-6 Antibody, R&D Systems™

Recombinant Monoclonal Antibody
$241.00 - $596.00
Specifications
Antigen | IL-6 |
---|---|
Clone | 1936R |
Dilution | Neutralization 8.00-80.0 ng/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-25 ug/mL |
Applications | Flow Cytometry, Neutralization |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB2061RSP
![]() |
R&D Systems
MAB2061RSP |
25 μg |
Each for $241.00
|
|
|||||
MAB2061R100
![]() |
R&D Systems
MAB2061R100 |
100 μg |
Each for $596.00
|
|
|||||
Description
IL-6 Monoclonal specifically detects IL-6 in Human samples. It is validated for Neutralization, Intracellular Staining by Flow Cytometry.Specifications
IL-6 | |
Neutralization 8.00-80.0 ng/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
P05231 | |
3569 | |
IgG2b | |
Protein A or G purified from cell culture supernatant | |
Detects human IL-6 in direct ELISAs. Does not cross-react with recombinant IL-6 from mouse, rat, or pig. |
1936R | |
Flow Cytometry, Neutralization | |
Unconjugated | |
Mouse | |
Human | |
B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A | |
IL6 | |
Primary | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title