Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AB206NA
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB206NA 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AB206NA Supplier R&D Systems Supplier No. AB206NA
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 5 publications

IL-6 Polyclonal specifically detects IL-6 in Human samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen IL-6
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Simple Western 50 μg/mL, Neutralization 0.05-0.15 μg/mL
Gene Accession No. P05231
Gene Alias B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A
Gene Symbols IL6
Host Species Goat
Immunogen E. coli-derived recombinant human IL-6 Pro29-Met212 Accession # P05231
Purification Method Protein A or G purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3569
Test Specificity Detects human IL-6 in direct ELISAs and Western blots. In these formats, less than 5% cross-reactivity with recombinant mouse IL-6, recombinant bovine IL-6, recombinant porcine IL-6, recombinant equine IL-6, recombinant feline IL-6, recombinant rat IL-6, recombinant cotton rat IL-6, recombinant rabbit IL-6, and recombinant viral IL-6 is observed.
Reconstitution Reconstitute at 1 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.