Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IL-6 Quantikine QuicKit ELISA

Catalog No. qk206
Click to view available options
Quantity:
1 Kit
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Assay Range 12.50 to 800 pg/mL
Assay Sensitivity 2.95 pg/mL
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Specificity Natural and Recombinant Human IL-6
For Use With (Application) ELISA
Gene ID (Entrez) 3569
Host Species Human
Quantity 1 Kit
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2 A
Target IL-6
Total Assay Time 80 minutes
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.