Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-6R alpha Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB227RS100UG
This item is not returnable.
View return policy
Description
IL-6R alpha Monoclonal specifically detects IL-6R alpha in Human samples. It is validated for Flow Cytometry.Specifications
IL-6R alpha | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
CD126, gp80, IL-6 R alpha, IL6Q, IL6R, IL6R alpha, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-6 R alpha Leu20-Asp339 Accession # P08887 | |
100 μg | |
3570 | |
Human | |
Supernatant |
Flow Cytometry | |
17506R | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P08887 | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human IL-6 R alpha in ELISAs and Western blots. In ELISAs, no cross-reactivity or interference was observed with recombinant human(rh) IL-1 alpha , recombinant mouse (rm) IL-1 alpha , rhIL-1 beta , rmIL-1 beta , rhIL-1ra, rhIL-2, rhIL-3, rmIL-3, rhIL-4, rmIL-4, rhIL-5, rmIL-5, rhIL-6, rmIL-6, rhIL-7, rmIL-7, rhIL-8, rhIL-9, rmIL-9, rhIL-10, or rhIL-11. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction