Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-6R alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB227500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB227500 500 μg
MAB227SP 25 μg
MAB227100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB227500 Supplier R&D Systems Supplier No. MAB227500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 18 publications

IL-6R alpha Monoclonal specifically detects IL-6R alpha in Human samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOF-ready, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen IL-6R alpha
Applications Western Blot, Flow Cytometry, Neutralization, CyTOF, ELISA
Classification Monoclonal
Clone 17506
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Neutralization 0.04-0.2 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P08887
Gene Alias CD126, gp80, IL-6 R alpha, IL6Q, IL6R, IL6R alpha, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor
Gene Symbols IL6R
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-6R alpha, Leu20-Asp339, Accession # P08887
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3570
Test Specificity Detects human IL-6R alpha in ELISAs and Western blots. In ELISAs, no cross-reactivity or interference was observed with recombinant human (rh) IL-1 alpha , recombinant mouse (rm) IL-1 alpha , rhIL-1 beta , rmIL-1 beta , rhIL-1ra, rhIL-2, rhIL-3, rmIL-3, r
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.