Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-6R alpha PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB227P025
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB227P025 25 Tests
FAB227P100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB227P025 Supplier R&D Systems Supplier No. FAB227P025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 1 publication

IL-6R alpha Monoclonal specifically detects IL-6R alpha in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-6R alpha
Applications Flow Cytometry
Classification Monoclonal
Clone 17506
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P08887
Gene Alias CD126, gp80, IL-6 R alpha, IL6Q, IL6R, IL6R alpha, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor
Gene Symbols IL6R
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-6 R alpha Leu20-Asp339 Accession # P08887
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3570
Test Specificity Detects human IL-6 R alpha in ELISAs and Western blots. In sandwich immunoassays, no cross-reactivity or interference was observed with recombinant human (rh) IL-1 alpha , recombinant mouse (rm) IL-1 alpha , rhIL-1 beta , rmIL-1 beta , rhIL-1ra, rhIL-2, rhIL-3, rmIL-3, rhIL-4, rmIL-4, rhIL-5, rmIL-5, rhIL-6, rmIL-6, rhIL-7, rmIL-7, rhIL-8, rhIL-9, rmIL-9, rhIL-10, or rhIL-11.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.