Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-7 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB207100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB207100 100 μg
MAB207SP 25 μg
MAB207500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB207100 Supplier R&D Systems Supplier No. MAB207100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 8 publications

IL-7 Monoclonal specifically detects IL-7 in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen IL-7
Applications Western Blot, Neutralization, ELISA
Classification Monoclonal
Clone 7417
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.4-0.8 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P13232
Gene Alias IL7, IL-7interleukin-7, interleukin 7, Lymphopoietin-1, PBGF
Gene Symbols IL7
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-7 Accession # P13232
Purification Method Protein A or G purified from ascites
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3574
Test Specificity Detects human IL-7 in Western blots. In ELISAs, does not cross-react with bovine FGF acidic, recombinant human (rh) G-CSF, rhGM-CSF, rhIL-1 alpha , rhIL-1 beta , rhIL-2, rhIL-3, rhIL-4, rhIL-6, recombinant mouse IL-7, rhIL-8, porcine PDGF, rhTNF-alpha , rhTNF-beta , human TGF-beta 1, or porcine TGF-beta 1.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Ascites
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.