Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-7 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$214.00 - $538.00
Specifications
Antigen | IL-7 |
---|---|
Clone | 7417R |
Dilution | Western Blot 1 μg/mL, Neutralization |
Applications | Western Blot, Neutralization |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB207RSP
![]() |
R&D Systems
MAB207RSP |
25 μg |
Each for $214.00
|
|
|||||
MAB207R100
![]() |
R&D Systems
MAB207R100 |
100 μg |
Each for $538.00
|
|
|||||
Description
IL-7 Monoclonal specifically detects IL-7 in Human samples. It is validated for Western Blot, Neutralization.Specifications
IL-7 | |
Western Blot 1 μg/mL, Neutralization | |
Monoclonal | |
Supernatant | |
Human | |
P13232 | |
3574 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
7417R | |
Western Blot, Neutralization | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
IL7, IL-7interleukin-7, interleukin 7, Lymphopoietin-1, PBGF | |
E. coli-derived recombinant human IL-7, Accession # P13232 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IL-7 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title