Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-7R alpha/CD127 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
$245.00 - $467.50
Specifications
Antigen | IL-7R alpha/CD127 |
---|---|
Clone | 40131 |
Dilution | Flow Cytometry 5 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB306G025
![]() |
R&D Systems
FAB306G025 |
25 Tests |
Each for $245.00
|
|
|||||
FAB306G100
![]() |
R&D Systems
FAB306G100 |
100 Tests |
Each for $467.50
|
|
|||||
Description
IL-7R alpha/CD127 Monoclonal specifically detects IL-7R alpha/CD127 in Human samples. It is validated for Flow Cytometry.Specifications
IL-7R alpha/CD127 | |
Flow Cytometry 5 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
3575 | |
Mouse myeloma cell line NS0-derived recombinant human IL-7 R alpha Extracellular domain | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
40131 | |
Flow Cytometry | |
Alexa Fluor 488 | |
Mouse | |
Human | |
CD127, CD127 antigen, CD127ILRA, CDW127, IL-7 R alpha, IL-7 receptor subunit alpha, IL7R, IL7R alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha | |
IL7R | |
IgG1 | |
Protein A or G purified from ascites | |
Detects human IL-7 R alpha in Western blots. In Western blots, approximately 5% cross-reactivity was observed with recombinant human (rh) IL-10 R, rhIL-2 R beta , rhIL-5 R alpha , and rhIL-6 R. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title