Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-7R alpha/CD127 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB306500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB306500 500 μg
MAB306SP 25 μg
MAB306100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB306500 Supplier R&D Systems Supplier No. MAB306500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 11 publications

IL-7R alpha/CD127 Monoclonal specifically detects IL-7R alpha/CD127 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.

Specifications

Antigen IL-7R alpha/CD127
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 40131
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias CD127, CD127 antigen, CD127ILRA, CDW127, IL-7 R alpha, IL-7 receptor subunit alpha, IL7R, IL7R alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha
Gene Symbols IL7R
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-7 R alpha Extracellular domain
Purification Method Protein A or G purified from ascites
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3575
Test Specificity Detects human IL-7 R alpha in Western blots. In Western blots, approximately 5% cross-reactivity was observed with recombinant human (rh) IL-10 R, rhIL-2 R beta , rhIL-5 R alpha , and rhIL-6 R.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Ascites
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.