Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-8/CXCL8 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF208NA
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF208NA 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF208NA Supplier R&D Systems Supplier No. AF208NA
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 9 publications

CXCL8/IL-8 Polyclonal specifically detects CXCL8/IL-8 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Neutralization, Immunocytochemistry.

Specifications

Antigen CXCL8/IL-8
Applications Western Blot, Immunohistochemistry, Neutralization, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Simple Western 25 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.1-0.5 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. P10145
Gene Alias 3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8, CXCL8SCYB8, Emoctakin, GCP1, GCP-1TSG-1, IL8, interleukin 8, K60, LAI, LECT, LUCT, LYNAP, MDNCF, MDNCFb-ENAP, member 8, MONAP, MONAPGCP1, NAF, NAP1, NAP-1NAP1, NCF, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor, TCF, TSG1
Gene Symbols IL8
Host Species Goat
Immunogen E. coli-derived recombinant human IL-8/CXCL8 Ser28-Ser99 Accession # P10145
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3576
Test Specificity Detects human IL-8/CXCL8 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant canine IL-8/CXCL8, recombinant feline IL-8/CXCL8 and recombinant porcine IL-8/CXCL8 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.