Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-8/CXCL8 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 32 publications
Supplier: R&D Systems BAF208
This item is not returnable.
View return policy
Description
CXCL8/IL-8 Polyclonal specifically detects CXCL8/IL-8 in Human samples. It is validated for Western Blot, Immunocytochemistry, ELISA Detection (Matched Antibody Pair).Specifications
CXCL8/IL-8 | |
Polyclonal | |
Biotin | |
P10145 | |
IL8 | |
E. coli-derived recombinant human IL-8/CXCL8 Ser28-Ser99 Accession # P10145 | |
50 μg | |
Primary | |
Detects human IL-8/CXCL8 in ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant porcine IL-8/CXCL8, recombinant rat (rr) CINC-2 alpha , and rrCINC-2 beta is observed. | |
Human | |
IgG |
Western Blot, Immunocytochemistry, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8, CXCL8SCYB8, Emoctakin, GCP1, GCP-1TSG-1, IL8, interleukin 8, K60, LAI, LECT, LUCT, LYNAP, MDNCF, MDNCFb-ENAP, member 8, MONAP, MONAPGCP1, NAF, NAP1, NAP-1NAP1, NCF, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor, TCF, TSG1 | |
Goat | |
Affinity Purified | |
RUO | |
3576 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction