Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IL-8/CXCL8 DuoSet ELISA
SDP

Catalog No. DY20805 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit (for 15 Plates)
1 Kit (for 5 Plates)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY20805 1 Kit (for 5 Plates)
DY208 1 Kit (for 15 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DY20805 Supplier R&D Systems™ Supplier No. DY20805

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Requires

DuoSet Ancillary Reagent Kit 2 - Catalog # DY008

OR

  • Wash Buffer - 0.05%Tween™ 20 in PBS, pH 7.2-7.4
  • Block Buffer - 1% BSA in PBS, pH 7.2-7.4
  • Reagent Diluent - 0.1% BSA, 0.05% Tween 20 in Tris-buffered Saline, pH 7.2-7.4, 0.2μm filtered
  • Substrate Solution - Catalog # DY999
  • Stop Solution - 2N H2SO4
  • Microplates - Catalog # DY990
  • ELISA Plate Sealers

Human IL-8/CXCL8 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IL-8/CXCL8 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human CXCL8/IL-8.

Specifications

Product Type Solid Phase Sandwich ELISA Development Kit
Quantity 1 Kit (for 5 Plates)
Species Human
Conjugate Biotin
Assay Sensitivity 31.20 to 2000 pg/mL
Assay Range 31.20 to 2000 pg/mL
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
For Use With (Application) ELISA
Gene ID (Entrez) 3576
Product Line R&D Systems
Synonym 3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8, CXCL8SCYB8, Emoctakin, GCP1, GCP-1TSG-1, IL8, interleukin 8, K60, LAI, LECT, LUCT, LYNAP, MDNCF, MDNCFb-ENAP, member 8, MONAP, MONAPGCP1, NAF, NAP1, NAP-1NAP1, NCF, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor, TCF, TSG1
Kit Contents Capture Antibody
Detection Antibody
Recombinant Standard
Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP)
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Research Area Chemokines
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.