Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-8/CXCL8 Fluorescein-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems IC208F
This item is not returnable.
View return policy
Description
CXCL8/IL-8 Monoclonal specifically detects CXCL8/IL-8 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
CXCL8/IL-8 | |
Monoclonal | |
Fluorescein | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8, CXCL8SCYB8, Emoctakin, GCP1, GCP-1TSG-1, IL8, interleukin 8, K60, LAI, LECT, LUCT, LYNAP, MDNCF, MDNCFb-ENAP, member 8, MONAP, MONAPGCP1, NAF, NAP1, NAP-1NAP1, NCF, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor, TCF, TSG1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3576 | |
Human | |
Purified |
Flow Cytometry | |
6217 | |
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
P10145 | |
IL8 | |
E. coli-derived recombinant human CXCL8/IL-8 Ser28-Ser99 Accession # P10145 | |
100 Tests | |
Primary | |
Detects human CXCL8/IL-8 in ELISAs and Western blots. In Western blots, this antibody shows 100% cross-reactivity with recombinant porcine CXCL8/IL-8 and no cross-reactivity with recombinant rat CXCL3/CINC-2 beta . | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction