Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IL10 (P22301) Recombinant Protein

Catalog No. 89948596 Shop All Abnova Corporation Products
Click to view available options
Quantity:
10 μg

Used for Func, SDS-PAGE

The protein encoded by this gene is a cytokine produced primarily by monocytes and to a lesser extent by lymphocytes. This cytokine has pleiotropic effects in immunoregulation and inflammation. It down-regulates the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. This cytokine can block NF-kappa B activity, and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice suggested the function of this cytokine as an essential immunoregulator in the intestinal tract. [provided by RefSeq]

Sequence: MSPGQGTQSENSCTHFPGNLPNmLRDLRDAFSRVKTFFQMKDQLDNLLLKESLLEDFKGYLGCQALSEMIQFYLEEVMPQAENQDPDIKAHVNSLGENLKTLRLRLRRCHRFLPCENKSKAVEQVKNAFNKLQEKGIYKAMSEFDIFINYIEAYMTMKIRN

Specifications

Accession Number P22301
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 3586
Molecular Weight (g/mol) 16.6kDa
Name IL10 (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Quantity 10 μg
Immunogen MSPGQGTQSENSCTHFPGNLPNMLRDLRDAFSRVKTFFQMKDQLDNLLLKESLLEDFKGYLGCQALSEMIQFYLEEVMPQAENQDPDIKAHVNSLGENLKTLRLRLRRCHRFLPCENKSKAVEQVKNAFNKLQEKGIYKAMSEFDIFINYIEAYMTMKIRN
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/μg
Gene Alias CSIF/IL-10/IL10A/MGC126450/MGC126451/TGIF
Common Name IL10
Gene Symbol IL10
Biological Activity The activity is determined by the dose-dependant proliferation of mouse MC-9 cells and is typically less than 0.5ng/mL.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.